Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41405

Sigma-Aldrich

(R)-3-Methyl-2-oxovaleric acid lithium salt hydrate

≥95.0% (TLC)

Synonym(s):

(R)-3-Methyl-2-oxopentanoic acid lithium salt, (R)-Ketoisoleucine lithium salt, Lithium (R)-3-methyl-2-oxovalerate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9LiO3 · xH2O
Molecular Weight:
136.07 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Product Name

(R)-3-Methyl-2-oxovaleric acid lithium salt hydrate, ≥95.0% (TLC)

assay

≥95.0% (TLC)

form

powder or crystals

optical activity

[α]/D -18.0±2.0°, c = 0.1 in hydrochloric acid

composition

carbon content, 52.96% (theory, anhydrous basis)
hydrogen content, 6.67% (theory, anhydrous basis)

color

white to off-white

storage temp.

2-8°C

SMILES string

[Li+].O.CC[C@@H](C)C(=O)C([O-])=O

InChI

1S/C6H10O3.Li.H2O/c1-3-4(2)5(7)6(8)9;;/h4H,3H2,1-2H3,(H,8,9);;1H2/q;+1;/p-1/t4-;;/m1../s1

InChI key

PTRNELNRMVEQEC-RZFWHQLPSA-M

Biochem/physiol Actions

3-Methyl-2-oxovaleric acid is a metabolite of isoleucine in human, animals and bacteria. Used as a clinical marker for maple syrup urine disease (MSUD)[1][2][3][4].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical approach in diagnosis of inherited metabolic diseases. Maple syrup urine disease (MSUD). Simultaneous analysis of metabolites in urine by enantioselective multidimensional capillary gas chromatography-mass spectrometry (enantio-MDGC-MS).
Podebrad, F., et al.
Journal of High Resolution Chromatography, 20, 355-362 (1997)
U Wendel et al.
Clinica chimica acta; international journal of clinical chemistry, 208(1-2), 85-91 (1992-06-15)
An enzymatic method for the separate measurement of both chiral 2-oxo-3-methylvaleric acid (OMV) compounds, (S)- and (R)-OMV, by NADH-dependent enantioselective amination using leucine dehydrogenase in the presence of a NADH regenerating system is described. This method allows the quantitative determination
Analytical approach in diagnosis of inherited metabolic diseases: Maple Syrup Urine Disease (MSUD).
Podebrad, F., et al.
GIT, 3, 15-16 (1999)
P Schadewaldt et al.
Journal of chromatography. B, Biomedical applications, 682(2), 209-218 (1996-07-12)
A sensitive method for the determination of S- and R-3-methyl-2-oxopentanoate enantiomers (KMV, alpha-keto-beta-methylvalerate) in physiological fluids suitable for isotope enrichment analysis is described: after extraction with acid, 2-oxo acids are separated from interfering amino acids by cation-exchange chromatography. Reductive amination
Synthesis and gas chromatography/mass spectrometry analysis of stereoisomers of 2-hydroxy-3-methylpentanoic acid.
O A Mamer
Methods in enzymology, 324, 3-10 (2000-09-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service