Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41180

Supelco

Micro particles, magnetic, protein A coated

1 μm particle size

Synonym(s):

Latex beads, magnetic, protein A coated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

particle size

1 μm

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service