Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40546

Sigma-Aldrich

3′-(6-FAM)-CPG

Synonym(s):

1-Dimethoxytrityloxy-3-(di-O-pivaloyl-fluorescein-6-carboxamidopropyl)-propyl-2-O-succinoyl-long chain alkylamino-CPG, 6-Carboxyfluorescein bound to controlled pore glass

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

storage temp.

−20°C

Application

6-Carboxyfluorescein bound to controlled pore glass (3′-(6-FAM)-CPG) may be used as a support for DNA and RNA oligo synthesis.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao-Hua Sun et al.
Biotechnology progress, 25(5), 1228-1235 (2009-07-16)
Pools of short synthetic oligonucleotides (oligos) are required in the multiplex and parallel DNA construction. Microarray technology provides a fast and economical mean for massive parallel synthesis of oligos. The method of oligo synthesis with the programmable microfluidic PicoArray could

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service