Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40487

Sigma-Aldrich

Sodium cis-homoaconitate tribasic

≥97.0% (HPLC)

Synonym(s):

(Z)-1-Butene-1,2,4-tricarboxylic acid trisodium salt, cis-Homoaconitic acid trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Na3O6
Molecular Weight:
254.08
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

assay

≥97.0% (HPLC)

form

powder

impurities

≤10% water

SMILES string

[Na+].[Na+].[Na+].[O-]C(=O)CC\C(=C\C([O-])=O)C([O-])=O

InChI

1S/C7H8O6.3Na/c8-5(9)2-1-4(7(12)13)3-6(10)11;;;/h3H,1-2H2,(H,8,9)(H,10,11)(H,12,13);;;/q;3*+1/p-3/b4-3-;;;

InChI key

LQKFCFKBZKHIQH-FGSKAQBVSA-K

application

Sodium cis-homoaconitate tribasic may be used as a substrate in studies of the biosynthesis of lysine in yeast.

Biochem/physiol Actions

cis-Homoaconitic acid is an intermediate in lysine biosynthesis[1], it is involved in the biosynthesis of coenzyme B in methanogenic Archaea[2].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haruhito Tsutsui et al.
Journal of proteome research, 9(8), 3912-3922 (2010-06-19)
The number of diabetic patients has recently been increasing worldwide. Thus, the discovery of potential diabetic biomarker(s), leading to the early detection and/or prevention of diabetes mellitus, is strongly required. The diagnosis of the prediabetic state in humans is a
Enzymatic formation of cis-homoaconitic acid, an intermediate in lysine biosynthesis in yeast.
M Strassman et al.
The Journal of biological chemistry, 241(22), 5401-5407 (1966-11-25)
M E Maragoudakis
Journal of bacteriology, 94(4), 1060-1065 (1967-10-01)
Homoaconitic acid, the second intermediate of the proposed pathway for lysine biosynthesis in yeast, is accumulated in the growth medium of a lysine-requiring mutant. This acid has been identified on paper and column chromatography by comparing it with authentic cis-homoaconitic
D M Howell et al.
Biochemistry, 37(28), 10108-10117 (1998-07-17)
The biochemistry of the 13 steps involved in the conversion of alpha-ketoglutarate and acetylCoA to alpha-ketosuberate, a precursor to the coenzymes coenzyme B (7-mercapto heptanoylthreonine phosphate) and biotin, has been established in Methanosarcina thermophila. These series of reactions begin with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service