Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40436

Sigma-Aldrich

Fluorescent red NIR 781 reactive

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H50N3NaO11S2
Molecular Weight:
859.98
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

suitability

in accordance for coupling to amines
in accordance for fluorescence
suitable for fluorescence

Quality Level

storage temp.

−20°C

SMILES string

[Na+].CCN(CC)c1ccc2c(cc(\C=C\C=C\C=C3\N(CCCS([O-])(=O)=O)c4ccc(cc4C3(C)CCCC(=O)ON5C(=O)CCC5=O)S([O-])(=O)=O)[o+]c2c1)C(C)(C)C

InChI

1S/C42H51N3O11S2.Na/c1-7-43(8-2)29-17-19-32-33(41(3,4)5)27-30(55-36(32)26-29)14-10-9-11-15-37-42(6,23-12-16-40(48)56-45-38(46)21-22-39(45)47)34-28-31(58(52,53)54)18-20-35(34)44(37)24-13-25-57(49,50)51;/h9-11,14-15,17-20,26-28H,7-8,12-13,16,21-25H2,1-6H3,(H-,49,50,51,52,53,54);/q;+1/p-1/b11-9+,14-10+,37-15+;

InChI key

DOZHKEVGVGFBGF-NNRGQRLSSA-M

Application

Fluorescent red 781 reactive is an amine conjugating fluorescent biolabel that can be used for covalent coupling to proteins and other biomolecules containing primary amino groups like amino modified DNA-oligomers and amino modified biotin. Fluorescent red 781 reactive is a new fluorescent label especially well suited for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required. Fluorescence NIR 781 reactive shows strong fluorescence. This holds also for its solid state emission.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service