Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40108

Sigma-Aldrich

Dimethyldecylphosphine oxide

≥98.0% (GC)

Synonym(s):

APO-10, Decyldimethylphosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9P(O)(CH3)2
CAS Number:
Molecular Weight:
218.32
Beilstein/REAXYS Number:
1930233
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

assay

≥98.0% (GC)

mol wt

micellar avg mol wt 28,597

aggregation number

131

CMC

4.6 mM (20-25°C)

λ

1 % in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.05

SMILES string

CCCCCCCCCCP(C)(C)=O

InChI

1S/C12H27OP/c1-4-5-6-7-8-9-10-11-12-14(2,3)13/h4-12H2,1-3H3

InChI key

GSVLCKASFMVUSW-UHFFFAOYSA-N

application

Non-ionic detergent useful for plasmid DNA isolation[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolf Huetteroth et al.
Frontiers in systems neuroscience, 4, 7-7 (2010-03-27)
DURING METAMORPHOSIS, THE TRANSITION FROM THE LARVA TO THE ADULT, THE INSECT BRAIN UNDERGOES CONSIDERABLE REMODELING: new neurons are integrated while larval neurons are remodeled or eliminated. One well acknowledged model to study metamorphic brain development is the sphinx moth
Plasmid DNA isolation utilizing a novel nonionic detergent.
M Altschuler et al.
BioTechniques, 17(3), 434-434 (1994-09-01)
Basil el Jundi et al.
PloS one, 6(11), e27855-e27855 (2011-11-25)
Mass migration of desert locusts is a common phenomenon in North Africa and the Middle East but how these insects navigate is still poorly understood. Laboratory studies suggest that locusts are able to exploit the sky polarization pattern as a
Yanliang Fan et al.
Scientific reports, 9(1), 3918-3918 (2019-03-10)
The anterior chamber of the eye (ACE) has emerged as a promising clinical islet transplantation site because of its multiple advantages over the conventional intra-hepatic portal site. This includes reduced surgical invasiveness and increased islet graft survival rate. It also
Yoshitaka Hamanaka et al.
PloS one, 7(7), e41109-e41109 (2012-07-31)
Butterflies have sophisticated color vision. While the spectral organization of the compound eye has been well characterized in the Japanese yellow swallowtail butterfly, Papilio xuthus, neural mechanisms underlying its color vision are largely unexplored. Towards a better understanding of signal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service