Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38737

Sigma-Aldrich

6,7-Dimethoxy-4-coumarinylacetic acid

BioReagent, suitable for fluorescence, ≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O6
CAS Number:
Molecular Weight:
264.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

assay

≥98.0% (TLC)

form

powder

solubility

DMSO: soluble
methanol: soluble

fluorescence

λex 342 nm; λem 418 nm in methanol

suitability

suitable for fluorescence

SMILES string

COc1cc2OC(=O)C=C(CC(O)=O)c2cc1OC

InChI

1S/C13H12O6/c1-17-10-5-8-7(3-12(14)15)4-13(16)19-9(8)6-11(10)18-2/h4-6H,3H2,1-2H3,(H,14,15)

Inchi Key

UKBVNOUQXDVDDX-UHFFFAOYSA-N

Application

6,7-Dimethoxy-4-coumarinyl acetic is a fluorescent molecule which may be used in fluorescent applications that use coumarinyl acetic acids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Márta Zarándi et al.
Journal of peptide science : an official publication of the European Peptide Society, 13(2), 94-99 (2006-11-15)
It has been proved that the principal component of senile plaques is aggregates of beta-amyloid peptide (Abeta) in cases of one of the most common forms of age-related neurodegenerative disorders, Alzheimer's disease (AD). Although the synthetic methods for the synthesis
L Fülöp et al.
Journal of peptide science : an official publication of the European Peptide Society, 7(8), 397-401 (2001-09-08)
Fluorescent cell analytical techniques require the incorporation of a fluorophore into the target molecule without causing a significant change in the native conformation. Many short peptides have a limited number of reactive groups that can be labeled without affecting the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service