Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38686

Sigma-Aldrich

D-Fructofuranose 2-phosphate potassium salt

≥90.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O9P · xK+
Molecular Weight:
260.14 (free acid basis)
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥90.0% (TLC)

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@H](O)[C@@H](CO)O[C@@]1(CO)OP(O)(O)=O

InChI

1S/C6H13O9P/c7-1-3-4(9)5(10)6(2-8,14-3)15-16(11,12)13/h3-5,7-10H,1-2H2,(H2,11,12,13)/t3-,4?,5+,6+/m1/s1

InChI key

PMTUDJVZIGZBIX-NQAPHZHOSA-N

Biochem/physiol Actions

Metabolite involved in the fructose and mannose metabolism, beta-D-Fructose 2-phosphate is produced from beta-D-Fructose 2,6-bisphosphate by the enzyme fructose-2,6- bisphosphate 6-phosphatase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Properties of D-fructose 1,2-cyclic phosphates and fructose 2-phosphates.
Taniguchi, H. and Nakamura, M.
Agricultural and Biological Chemistry, 36, 2373-2380 (1972)
Hydrolysis and Isomerization of Sugar Phosphates and Carbohydrate Phosphodiesters.
Mikkola, S.
Current Organic Chemistry, 17, 1525-1544 (2013)
C Purwin et al.
European journal of biochemistry, 165(3), 543-545 (1987-06-15)
Using comparative ion-exchange chromatography on Dowex 1X4, the product of dephosphorylation of fructose 2,6-bisphosphate with purified yeast fructose-2,6-bisphosphate 6-phosphohydrolase, was shown to be identical to the furanose form of fructose 2-phosphate prepared by chemical synthesis according to Pontis and Fischer
SYNTHESIS OF D-FRUCTOPYRANOSE 2-PHOSPHATE AND D-FRUCTOFURANOSE 2-PHOSPHATE.
H G PONTIS et al.
The Biochemical journal, 89, 452-459 (1963-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service