Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38605

Sigma-Aldrich

9,10-Dimethoxy-2-anthracenesulfonic acid sodium salt

for fluorescence, ≥96.0% (HPLC)

Synonym(s):

DAS-Na, Na-DAS, Sodium 9,10-dimethoxyanthracene-2-sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NaO5S
CAS Number:
Molecular Weight:
340.33
Beilstein/REAXYS Number:
4285639
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

grade

for fluorescence

assay

≥96.0% (HPLC)

suitability

suitable for fluorescence

SMILES string

[Na+].COc1c2ccccc2c(OC)c3cc(ccc13)S([O-])(=O)=O

InChI

1S/C16H14O5S.Na/c1-20-15-11-5-3-4-6-12(11)16(21-2)14-9-10(22(17,18)19)7-8-13(14)15;/h3-9H,1-2H3,(H,17,18,19);/q;+1/p-1

InChI key

CBGQZXMEEWCJQD-UHFFFAOYSA-M

Application

9,10-Dimethoxy-2-anthracenesulfonic acid is used as a fluorometric ion-pair reagent for post-column determination of amine containing compounds such as alkaloids [1][2][3]

Other Notes

Reagent for the determination of amines (e.g. alkaloids) using the fluorometric ion-pair technique as a post-column reactor of HPLC[1][2][3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Abdine et al.
Die Pharmazie, 60(4), 265-268 (2005-05-11)
A simple and sensitive spectrofluorometric method was developed for the quantitative determination of some beta-blockers, namely arotinolol, atenolol and labetalol as hydrochloride salts. The method is based on the reaction of these drugs as n-electron donors with the fluorogenic reagent
J.M. Huen et al.
Resolut. Chromatogr. Commun., 3, 154-154 (1979)
K Kümmerer et al.
Journal of chromatography. A, 774(1-2), 281-286 (1997-07-11)
A highly reproducible and specific method for the analysis of the quaternary ammonium compound, benzalkonium chloride, in effluents from European hospitals is presented. Benzalkonium chloride was extracted with end-capped RP-18 solid-phase cartridges and was selectively eluted. The resulting solution was
D. Westerland et al.
Analytica Chimica Acta, 67, 89-89 (1973)
J.C. Gfeller et al.
Journal of Chromatography A, 172, 141-141 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service