Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38261

Sigma-Aldrich

3-Sulfo-N-succinimidyl benzoate-(phenyl-13C6) sodium salt

≥95% (HPLC)

Synonym(s):

Benzoic acid (3-sulfo-N-succinimidyl) ester-(phenyl-13C6) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C5H8NNaO7S
CAS Number:
Molecular Weight:
327.19
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.25

assay

≥95% (HPLC)

storage temp.

2-8°C

SMILES string

[Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)[13c]2[13cH][13cH][13cH][13cH][13cH]2)C1=O

InChI

1S/C11H9NO7S.Na/c13-9-6-8(20(16,17)18)10(14)12(9)19-11(15)7-4-2-1-3-5-7;/h1-5,8H,6H2,(H,16,17,18);/q;+1/p-1/i1+1,2+1,3+1,4+1,5+1,7+1;

InChI key

MDKFDOUBUQSEDS-JLLOWZHGSA-M

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of Coenzyme A Esters in the Metabolism of Benzoate and Cyclohexanecarboxylate by Rhodopseudomonas palustris.
Georgina N. Hutber
Microbiology, 129, 2413-2420 (1983)
J V Staros
Biochemistry, 21(17), 3950-3955 (1982-08-17)
We have synthesized and characterized N-hydroxysulfosuccinimide, a new hydrophilic ligand for the preparation of active esters. We have incorporated this ligand into two new protein cross-linking reagents, 3,3'-dithiobis(sulfosuccinimidyl propionate) and bis(sulfosuccinimidyl) suberate. In experiments with rabbit muscle aldolase, it is
Alexandre Panchaud et al.
Molecular & cellular proteomics : MCP, 7(4), 800-812 (2007-12-18)
Identification and relative quantification of hundreds to thousands of proteins within complex biological samples have become realistic with the emergence of stable isotope labeling in combination with high throughput mass spectrometry. However, all current chemical approaches target a single amino

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service