Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

381S

Sigma-Aldrich

TIA-1 Positive Control Slides

suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.24
Product 381S is not currently sold in your country. Contact Technical Service

packaging

pkg of 5 slides

Quality Level

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

shipped in

ambient

storage temp.

2-8°C

General description

TIA-1 positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-TIA-1 antibody.

Physical form

Paraffin embedded tissue section mounted on microscope slide

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camilla Pandolfi et al.
Tree physiology, 37(3), 380-388 (2017-03-25)
Saline soils are highly heterogeneous in time and space, and this is a critical factor influencing plant physiology and productivity. Temporal changes in soil salinity can alter plant responses to salinity, and pre-treating plants with low NaCl concentrations has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service