Key Documents
37200
Supelco
Sulfite testing paper
for detection of sulfite, sulfurous acid and sulfur dioxide
About This Item
UNSPSC Code:
12352103
quality
for detection of sulfite, sulfurous acid and sulfur dioxide
Quality Level
suitability
corresponds for sensitivity to SO3
application(s)
general analytical
Application
It may be used to identify sulphite in urine in an attempt to diagnose sulphite oxidase deficiency.[1]
Packaging
box with 100 strips
Other Notes
this product will be discontinued, please find the alternative product here :
110013
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Attempt at therapy in sulphite oxidase deficiency.
P Tardy et al.
Journal of inherited metabolic disease, 12(1), 94-95 (1989-01-01)