Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36911

Supelco

α,α,α-Trichlorotoluene

analytical standard

Synonym(s):

(Trichloromethyl)benzene, Benzotrichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CCl3
CAS Number:
Molecular Weight:
195.47
Beilstein/REAXYS Number:
508152
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor density

6.77 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

autoignition temp.

411 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.557 (lit.)

bp

219-223 °C/760 mmHg (lit.)
90-91 °C/15 mmHg (lit.)

mp

−7.5-−7 °C (lit.)

density

1.38 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

ClC(Cl)(Cl)c1ccccc1

InChI

1S/C7H5Cl3/c8-7(9,10)6-4-2-1-3-5-6/h1-5H

InChI key

XEMRAKSQROQPBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

228.2 °F - closed cup

flash_point_c

109 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Wong
American journal of industrial medicine, 14(4), 417-431 (1988-01-01)
The cohort of this historical prospective mortality study consisted of 697 male employees at a chlorination plant. A majority of the cohort was potentially exposed to benzotrichloride, benzyl chloride, benzoyl chloride, and other related chemicals. The mortality experience of the
Benzotrichloride.
Report on carcinogens : carcinogen profiles, 11, III31-III32 (2004-01-01)
Tamar Kohn et al.
Environmental science & technology, 40(13), 4253-4260 (2006-07-22)
Cross-correlations of rate constants between a system of interest and a better-defined one have become popular as a tool in studying transformations of organic pollutants. A slope of unity (if the correlation is conducted on a log-log basis) in such
M Govindarajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 94, 53-64 (2012-04-21)
FT-IR and FT-Raman spectra of α,α,α-trichlorotoluene have been recorded and analyzed. The geometry, fundamental vibrational frequencies are interpreted with the aid of structure optimizations and normal coordinate force field calculations based on density functional theory (DFT) B3LYP/6-311++G(d,p) method and a
Benzotrichloride.
Report on carcinogens : carcinogen profiles, 10, 30-31 (2004-08-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service