Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36659

Supelco

α-Endosulfan solution

new

100 μg/mL in hexane, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6Cl6O3S
CAS Number:
Molecular Weight:
406.93
Beilstein/REAXYS Number:
2950316
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in hexane

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C2(Cl)C3COS(=O)OCC3C1(Cl)C2(Cl)Cl

InChI

1S/C9H6Cl6O3S/c10-5-6(11)8(13)4-2-18-19(16)17-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3-,4+,7-,8+,19+

InChI key

RDYMFSUJUZBWLH-BEJSPLBDSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-14.8 °F - closed cup

flash_point_c

-26 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service