Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36535

Sigma-Aldrich

Atto Rho6G alkyne

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

product line

BioReagent

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 535 nm; λem 560 nm±10 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Rho6G is a new rhodamine dye, based on the well-known laser dye Rhodamine 6G. The new label is functionalized for coupling to biomolecules such as DNA, RNA or proteins. Atto Rho6G shows strong absorption, extraordinary high fluorescence quantum yield, high thermal and photo-stability, and very little triplet formation. The dye is highly suitable for single-molecule detection applications and high-resolution microscopy.
After coupling to a substrate Atto Rho6G carries a net electric charge of +1. The label is moderately hydrophilic.
The alkyne modification is used in the Huisgen reaction (“Click Chemistry“).

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service