Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3500-096-K

Sigma-Aldrich

Cultrex 3-D Spheroid Cell Invasion Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

shipped in

dry ice

storage temp.

−70°C

General description

The need for more complete and physiologically predictive cancer invasion models has driven the development of the 3-D Culture 96 Well BME Cell Invasion Assay. There is growing evidence that tumor cell aggregates or spheroids are more representative of tumors in vivo, and they exhibit several physiological traits including similar morphology, the formation of cell-cell bonds, decreased proliferation rates, increased cell survival, tumor dormancy, and a hypoxic core. Applying this model to in a 3-D culture invasion assay provides a more physiological approach for assessing tumor invasion and providing a visual component that can be quantitated through image analysis.

Other Notes

Storage Conditions and Product Stability:
Components are stored at room temperature and -80°C. Strip seals are provided to preserve sterility of unused wells from the 96 Well, round bottom plate for subsequent experiments.

Legal Information

6538 is a trademark of American Type Culture Collection

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service