Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

332A-1

Sigma-Aldrich

Somatostatin Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (332A-14)
vial of 0.5 mL concentrate (332A-15)
bottle of 1.0 mL predilute (332A-17)
vial of 1.0 mL concentrate (332A-16)
bottle of 7.0 mL predilute (332A-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200-1:1000

control

pancreas

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... SST(6750)

General description

Anti-Somatostatin is a useful marker of D-cells of pancreatic islet cells. D-cells identification can be used to identify hyperplasia of the pancreatic islets as well as islet cell tumors such as somatostatinomas. Somatostain can also be found in gastrointestinal and bronchopulmonary endocrine cells, thymic endocrine cells, and thyroid C cells. Somatostatin suppresses gastric acid secretion, gallbladder contractions, and pancreatic enzyme secretion.

Quality


IVD

IVD

IVD

RUO

Linkage

Somatostatin Positive Control Slides , Product No. 332S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Somers et al.
Gastroenterology, 85(5), 1192-1198 (1983-11-01)
A duodenal somatostatinoma is described in a patient suffering from a tumoral obstruction of the bile duct. The diagnosis was based upon histologic and immunocytochemical characterization of tumoral fragments and later confirmed by immunochemical analysis of liver metastases. Basal peripheral
M Quartu et al.
Journal of chemical neuroanatomy, 6(2), 79-99 (1993-03-01)
The occurrence of vasoactive intestinal polypeptide (VIP), peptide histidine-isoleucine (PHI), calcitonin gene-related peptide (CGRP), substance P (SP), somatostatin (SOM), galanin (GAL) and enkephalins (ENK) is studied in the human celiac/superior mesenteric ganglionic complex of pre- and full-term newborns, and adult
G Fried et al.
Cancer, 74(1), 142-151 (1994-07-01)
A case of a renin-producing paraganglioma of adrenal origin with metastases to the retroperitoneal area, paravaginal area, and the ovary is reported with immunohistochemical findings indicating expression of multiple neuropeptide immunoreactivities. The patient was 23 years old at the time
G Chejfec et al.
Ultrastructural pathology, 16(5), 537-545 (1992-09-01)
Upper gastrointestinal tract neuroendocrine tumors producing predominantly somatostatin have thus far been described only in the duodenum; their characteristic features include the frequent presence of psammoma bodies (psammomatous somatostinomas), and the association with von Recklinghausen's neurofibromatosis. Gastric neuroendocrine tumors, on
Types of pancreatic islet cells and their immunocytochemical identification
Erlandsen, SL
The Pancreas, 140-155 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service