Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31437

Millipore

DEV Lactose Peptone Broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.74

sterility

non-sterile

shelf life

limited shelf life, expiry date on the label

composition

bromocresol purple, 0.01 g/L
lactose, 10.0 g/L
meat peptone, 10.0 g/L
sodium chloride, 5.0 g/L

final pH

7.0±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

nonselective and differential for Escherichia coli
nonselective and differential for bacteria (General Media)
nonselective and differential for coliforms

Application

For the enrichment and titre determination of coliform bacteria acc. to the "Deutschen Einheitsverfahren" for the bacteriological control of drinking water.

Other Notes

Supervision of the hygienic quality of bathing water by detection of E. coli after a 7-day salt water stress period[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kolbeck et al.
Zentralblatt fur Hygiene und Umweltmedizin = International journal of hygiene and environmental medicine, 193(3), 224-229 (1992-10-01)
Fecal coliform bacteria (E. coli) are the most important parameter when monitoring the hygienic quality of bathing water according to EC guidelines. Among other things the number of E. coli is clearly dependent on the length of exposure of the

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service