Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31432

Millipore

DEV ENDO Agar

suitable for microbiology

Synonym(s):

Lactose Fuchsin Sulfite Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

agar, 20.0 g/L
fuchsin, 0.5 g/L
lactose, 10.0 g/L
meat extract, 10.0 g/L
meat peptone, 10.0 g/L
sodium chloride, 5.0 g/L
sodium sulfite, 2.5 g/L

final pH

7.3±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

selective and differential for Escherichia coli
selective and differential for coliforms

Application

In accordance with DIN-Norm 38411 for the examination of water
Selective agar for the isolation and differentiation of E. coli acc. to the "Deutschen Einheitsverfahren" for the bacteriological control of drinking water.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service