Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30516

Sigma-Aldrich

Trehalose 6-tetradecanoate

≥95.0% (GC)

Synonym(s):

α,α-Trehalose 6-myristate, α-D-Glucopyranosyl-α-D-glucopyranoside 6-tetradecanoate, 6-O-Myristoyl-α,α-trehalose, Trehalose 6-myristate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H48O12
CAS Number:
Molecular Weight:
552.65
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

assay

≥95.0% (GC)

form

powder

CMC

0.012 mM

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1O[C@H]2O[C@H](COC(CCCCCCCCCCCCC)=O)[C@@H](O)[C@H](O)[C@H]2O

InChI

1S/C26H48O12/c1-2-3-4-5-6-7-8-9-10-11-12-13-18(28)35-15-17-20(30)22(32)24(34)26(37-17)38-25-23(33)21(31)19(29)16(14-27)36-25/h16-17,19-27,29-34H,2-15H2,1H3/t16?,17?,19-,20-,21?,22?,23+,24+,25-,26-/m1/s1

InChI key

LRXBVTKVELRWDT-AULNSQLGSA-N

Application

Trehalose is commonly used as a stabilizer and protective material for biomaterial compounds. Because of these characteristics, Trehalose is introduced as a hydrophilic part. Due to these properties of Trehalose, protein degeneration is expected to be prevented with Trehalose detergents. Therefore, Trehalose detergents may keep the function of the protein and be utilized in the field of proteomics research.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Displacer Chemistry on Displacer Efficacy for a Sugar-Based Anion Exchange Displacer Library.
Liu, J., et al.
Industrial & Engineering Chemistry Research, 45, 9107-9114 (2006)
Chemical and biochemical studies on carbohydrate esters. V. Anti Ehrlich ascites tumor effect and chromatographic behaviors of fatty acyl monoesters of sucrose and trehalose.
Y Nishikawa et al.
Chemical & pharmaceutical bulletin, 25(7), 1717-1724 (1977-07-01)
Surface activities of monoacyl trehaloses in aqueous solution.
Chen, J., et al.
Food Sci. Technol., 40, 412-417 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service