Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278R-1

Sigma-Aldrich

Lysozyme (EP134) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

rabbit

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP134, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (278R-17)
bottle of 7.0 mL predilute (278R-18)
vial of 0.1 mL concentrate (278R-14)
vial of 0.5 mL concentrate (278R-15)
vial of 1.0 mL concentrate (278R-16)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

Anti-lysozyme stains myeloid cells, histiocytes, granulocytes, macrophages, and monocytes. It is an important marker that may demonstrate the myeloid or monocytic nature of acute leukemia. The restrictive nature of anti-lysozyme staining suggests that lysozyme may be synthesized predominantly in reactive histiocytes rather than in resting, unstimulated phagocytes. Anti-lysozyme may aid in the identification of histiocytic neoplasias, large lymphocytes, and classifying lymphoproliferative disorders.1-3

Quality


IVD

IVD

IVD

RUO

Linkage

Lysozyme Positive Control Slides, Product No. 278S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Delaflor-Weiss et al.
Acta cytologica, 43(6), 1124-1130 (1999-12-01)
Granulocytic sarcoma of the uterine cervix is an unusual manifestation of acute myeloid leukemia, representing soft tissue masses of leukemic myeloblasts. An often misdiagnosed entity, it is often confused with other inflammatory or neoplastic conditions, including large cell lymphoma. A
L Krugliak et al.
American journal of hematology, 21(1), 99-109 (1986-01-01)
Blood and bone marrow samples from 20 individuals with reactive conditions and 26 cases of acute and chronic myeloid leukemias were tested for the presence of lysozyme, alpha-1-antitrypsin (alpha-1-AT), and alpha-1-antichymotrypsin (alpha-1-ACT). We compared the reactivity of samples in smears

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service