26746
Cholesterol Oxidase from Nocardia erythropolis
in 1 M ammonium sulfate solution, pH 6, solution (slightly hazy), ≥15 U/mL
About This Item
Recommended Products
form
solution (slightly hazy)
quality
in 1 M ammonium sulfate solution, pH 6
concentration
≥15 U/mL
color
pale yellow
storage temp.
2-8°C
Unit Definition
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service