Key Documents
2503S
Millipore
BIOEAZE EZ Top and Aseptic Connecting Devices
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available. Contact Technical Service
description
Designed for the Aseptic Transfer of Liquids into or from 500 mL or 1000 mL Bottles with the Terumo Aseptic Connecting Device (ACD)
sterility
sterile; γ-irradiated
General description
Fits 500 mL or 1 L Bottle with 082 C-Flex® Stopper and 38 mm Cap with 1 x 2 ft. length of 1/8" x 1/4" 082 Clear C-Flex® Tubing with PInch Clamp and Plug
Packaging
Double Polybagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)