Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21050

Sigma-Aldrich

E-Toxate Reagent

sufficient for 50 tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

dry concentrate

Quality Level

usage

sufficient for 50 tests

IVD

not for in vitro diagnostic use

sensitivity measuring range

0.05-0.1 EU/mL

storage temp.

−20°C

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC

comparable product

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Boey et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 67(5), 351-360 (2019-01-10)
Modern electron microscopy offers a wide variety of tools to investigate the ultrastructural organization of cells and tissues and to accurately pinpoint intracellular localizations of macromolecules of interest. New volumetric electron microscopy techniques and new instrumentation provide unique opportunities for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service