Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

21020

Sigma-Aldrich

E-Toxate Reagent

sufficient for 20 tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

dry concentrate

Quality Level

usage

sufficient for 20 tests

IVD

not for in vitro diagnostic use

sensitivity measuring range

0.05-0.1 EU/mL

storage temp.

−20°C

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugen Brailoiu et al.
Cell reports, 26(2), 330-337 (2019-01-10)
Sigma-1 receptors (Sig-1Rs) are integral ER membrane proteins. They bind diverse ligands, including psychoactive drugs, and regulate many signaling proteins, including the inositol 1,4,5-trisphosphate receptors (IP3Rs) that release Ca2+ from the ER. The endogenous ligands of Sig-1Rs are unknown. Phospholipase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service