Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

201201

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate

~95%

Synonym(s):

β-NADPH, 2′-NADPH hydrate, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N7Na4O17P3 · xH2O
CAS Number:
Molecular Weight:
833.35 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

assay

~95%

form

solid

packaging

preweighed vial of 1 mg

SMILES string

[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@H]2O[C@@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@H](O)[C@@H]2O

InChI

1S/C21H30N7O17P3.4Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;;;/h1,3-4,7-8,10-11,13-16,20-21,29-31H,2,5-6H2,(H2,23,32)(H,36,37)(H,38,39)(H2,22,24,25)(H2,33,34,35);;;;/q;4*+1/p-4/t10-,11+,13-,14+,15-,16+,20-,21+;;;;/m0..../s1

InChI key

WYWWVJHQDVCHKF-MPUNMZHWSA-J

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

NADPH is an electron donor and a cofactor for many redox enzymes including nitric oxide synthetase.

Preparation Note

Chemically reduced

Other Notes

Packaged based on solid weight.

comparable product

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service