Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19551

Sigma-Aldrich

1-Ethyl-3,3,3′,3′-tetramethyl-1′-[5-(N-succinimidyloxycarbonyl)pentyl]indocarbocyanine iodide

suitable for fluorescence

Synonym(s):

2-[3-[1-[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-1,3-dihydro-3,3-dimethyl-2H-indol-2-ylidene]-1-propen-1-yl]-1-ethyl-3,3-dimethyl-3H-indolium iodide, IC3-OSU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H42IN3O4
CAS Number:
Molecular Weight:
695.63
UNSPSC Code:
12352200
PubChem Substance ID:

solubility

DMSO: 0.5 mg/mL, clear, slightly red to deep red

fluorescence

λex 550 nm; λem 570.0 nm±5 nm

suitability

suitable for fluorescence

Storage temp.

−20°C

General description

IC3 and IC5 are indocyanine dyes that can react with amine groups on proteins, peptides, and amine-modified oligonucleotides. Because neither IC3 nor IC5 has a sulfonate group in its structure, water solubility of the molecules is very low. An organic solvent is required to dissolve the compound. Over-labeling of proteins with these compounds may precipitate the labeled protein from aqueous solutions as a result of the poor water solubility. However, because no negative charge is added to the protein, the total number of charges of the modified protein is the same before and after labeling with these compounds. The IC dye package contains 100 μg dye. This amount is sufficient to modify 1-2 mg proteins or more depending on the purpose of the downstream experiment.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service