Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19546

Supelco

pIMAGO-biotin Phosphoprotein Detection Kit

for Microplate, Fluor 680 detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116134

manufacturer/tradename

Tymora Analytical Operations, LLC

detection method

fluorometric

storage temp.

2-8°C

General description

pIMAGO is a universal phosphoprotein detection technology that enables sensitive and specific recognition of phosphorylated molecules. Unlike phospho-antibodies, the binding is not biased by amino acid sequence, and therefore can be used for detection of any phosphorylation event on any
protein site. pIMAGO detection protocol resembles a simple ELISA procedure and can be easily incorporated by any laboratory. Due to its small size, pIMAGO can be multiplexed with antibodies for simultaneous detection of phosphorylation and total protein amount.

Legal Information

pIMAGO is a trademark of Tymora Analytical Operations, LLC

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service