Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19099

Sigma-Aldrich

Fluorescent Red 635 maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H52N4O7S
Molecular Weight:
780.97
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

powder

concentration

≥60% (degree of coupling)

fluorescence

λex 640 nm±5 nm; λem 669 nm±5 nm in 0.1 M phosphate pH 7.0

suitability

in accordance for fluorescence

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cc(\C=C\C=C2\N(CCCCCC(=O)NCCN3C(=O)C=CC3=O)c4ccc(cc4C2(C)C)S([O-])(=O)=O)c5cc6CCCN7CCCc(c67)c5[o+]1

InChI

1S/C44H52N4O7S/c1-43(2,3)37-27-29(33-26-30-13-10-22-46-23-11-14-32(41(30)46)42(33)55-37)12-9-15-36-44(4,5)34-28-31(56(52,53)54)17-18-35(34)47(36)24-8-6-7-16-38(49)45-21-25-48-39(50)19-20-40(48)51/h9,12,15,17-20,26-28H,6-8,10-11,13-14,16,21-25H2,1-5H3,(H-,45,49,52,53,54)/b12-9+,36-15+

InChI key

FMQYCGLOBYBNIH-ZUCLULJASA-N

Application

Fluorescent orange 635 maleimide is a thiol reactive fluorescent biolabel that can be used for covalent coupling to proteins, glutathione and other biomolecules containing sulfhydryl groups.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service