Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18114

Sigma-Aldrich

Fluorescent Orange 555 maleimide

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H47N5O7
Molecular Weight:
721.84
MDL number:
UNSPSC Code:
12352200

form

solid

fluorescence

λex 561 nm; λem 584 nm in DMF (250μg/mL)

suitability

suitable for fluorescence

storage temp.

2-8°C

Application

Fluorescent orange 555 maleimide is an thiol reactive fluorescent biolabel that can be used for covalent coupling Fluorescent orange 555 to proteins and other biomolecules containing sulfhydryl groups.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service