Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18047

Sigma-Aldrich

Bromophenol blue-Xylenecyanol solid mixture

powder (lyophilized)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

form

powder (lyophilized)

pH

~8.3

SMILES string

[Na+].Oc1c(Br)cc(cc1Br)C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4.CCNc5ccc(cc5C)\C(=C6/C=CC(\C=C6)=N/CC)c7ccc(cc7S(O)(=O)=O)S([O-])(=O)=O

InChI

1S/C24H26N2O6S2.C19H10Br4O5S.Na/c1-4-25-19-9-6-17(7-10-19)24(18-8-13-22(26-5-2)16(3)14-18)21-12-11-20(33(27,28)29)15-23(21)34(30,31)32;20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19;/h6-15,26H,4-5H2,1-3H3,(H,27,28,29)(H,30,31,32);1-8,24-25H;/q;;+1/p-1/b24-17-,25-19+;;

InChI key

ONUYXLWQPRRMIM-ODTFGHENSA-M

Application

Tracking dye for DNA sequencing gel electrophoresis

Reconstitution

when reconstituted to 5 mL, solution contains 0.5% Bromophenol Blue and 0.5% Xylenecyanol FF in Tris-borate-EDTA buffer

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Florencia Bambace et al.
Food research international (Ottawa, Ont.), 122, 653-660 (2019-06-24)
Minimally processed fruits are an alternative to dairy products to deliver probiotics. Bio-protection against several factors that affect their viability has been proposed in the food industry. In this study, probiotic Lactobacillus rhamnosus CECT 8361 was added to alginate-based coatings

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service