Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

174R-1

Sigma-Aldrich

CD74 (EP167) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Product 174R-1 is not currently sold in your country. Contact Technical Service

biological source

rabbit

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP167, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (174R-14)
vial of 0.1 mL concentrate Research Use Only (174R-14-RUO)
vial of 0.5 mL concentrate (174R-15)
vial of 1.0 mL concentrate (174R-16)
vial of 1.0 mL concentrate Research Use Only (174R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (174R-17-RUO)
vial of 1.0 mL pre-dilute ready-to-use (174R-17)
vial of 7.0 mL pre-dilute ready-to-use (174R-18)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (174R-18-RUO)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200 (concentrated)

isotype

IgG

control

lymph node, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, membranous

Gene Information

human ... CD74(972)

General description

The CD74 antigen represents a membrane-bound subunit of the MHC Class II-associated invariant chain that is encoded by the gene located on Chromosome 5. The antibody recognizes nuclear and cytoplasmic antigens of MW 35 kD and 31 kD respectively. Anti-CD74 stains predominantly germinal center lymphocytes and those in the mantle cell region of the lymph node. It stains the cell membrane but a paranuclear globular labeling is also noted. Plasma cells are not labeled. Thymocytes are negative but dendritic cells are often labeled by anti-CD74. Anti-CD74 labels about 90% of low grade B-cell lymphomas and 20% of low grade T-cell lymphomas. In high-grade lymphomas the discriminatory power (B-cell vs. T-cell) is diminished. Reed-Sternberg cells are also labeled in approximately 60% of cases.1

Quality


IVD

IVD

IVD

RUO

Linkage

CD74 Positive Control Slides, Product No. 174S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manual of Diagnostic Antibodies for Immunohistology (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service