Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17088

Sigma-Aldrich

Indo-1FF-AM

suitable for fluorescence, ≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H47F2N3O22
Molecular Weight:
1031.87
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

assay

≥95% (TLC)

solubility

DMSO: soluble

fluorescence

λex 350 nm; λem 435 nm in 0.1 M Tris pH 8.0, esterase; 10 mM Ca2+

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)c1ccc(cc1OCCOc2c(F)c(F)ccc2N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)-c3c[nH]c4cc(ccc34)C(=O)OCOC(C)=O

InChI

1S/C46H47F2N3O22/c1-26(52)64-21-69-40(57)17-50(18-41(58)70-22-65-27(2)53)37-10-7-31(34-16-49-36-14-32(6-8-33(34)36)46(61)73-25-68-30(5)56)15-39(37)62-12-13-63-45-38(11-9-35(47)44(45)48)51(19-42(59)71-23-66-28(3)54)20-43(60)72-24-67-29(4)55/h6-11,14-16,49H,12-13,17-25H2,1-5H3

InChI key

VSJCENCIDAGWSX-UHFFFAOYSA-N

Features and Benefits

Pale yellow green solid. Cell permeable acetoxymethyl ester (AM) derivative of Indo-1FF with spectral properties similar to the parent compound. A ratiometric dye with the same fluorescent properties and similar applications as Indo-1, but has a lower affinity for calcium, and is insensitive to magnesium[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganitkevich VYa
Cell calcium, 23(5), 313-322 (1998-07-29)
A low-affinity fluorescent Ca2+ indicator Indo-1FF was used to measure cytoplasmic Ca2+ increments in single smooth muscle cells isolated from the urinary bladder of the guinea-pig. The in vitro Kd of Indo-1FF for Ca2+ measured at the microscope stage was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service