Key Documents
1654R
Millipore
BIOEAZE Rigging Sets
About This Item
UNSPSC Code:
12352200
description
Designed for the Aseptic Transfer of Liquids into or out of Bottles or Bags, Double Polybagged
sterility
sterile; γ-irradiated
General description
9" Rigid Polypropylene Tube with 1 x 2 ft. length of Size 15 Opaque 001 C-Flex® Tubing and 1/4" Male Quick Connect with Cap
Packaging
Double Polybagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)