Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16441

Sigma-Aldrich

Lectin from Triticus vulgaris-Atto 488 conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

description

carbohydrate specificity (GlcNAc)2, NeuNAc

form

powder

fluorescence

λex 501 nm; λem 523 nm in PBS

storage temp.

−20°C

General description

Lectins are ubiquitous proteins or glycoproteins with a high and specific affinity to sugar residues. They allow a sensitive detection of cellular carbohydrates, revealing subtle alteration in glycosylation between otherwise indistinguishable cells.
Atto-dyes have very bright fluorescent signals and high photostability, which enable a direct one-step tissue-binding protocol. Time-consuming multistage amplification procedures are not required for Atto-dye lectin conjugates.

find more information here

Analysis Note

free of unconjugated dye

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleksandr Kolyvushko et al.
Microorganisms, 8(7) (2020-07-11)
Viruses utilize host cell signaling to facilitate productive infection. Equine herpesvirus type 1 (EHV-1) has been shown to activate Ca2+ release and phospholipase C upon contact with α4β1 integrins on the cell surface. Signaling molecules, including small GTPases, have been
Flexible Microdomain Specific Staining of Block Copolymers for 3D Optical Nanoscopy.
Ullal, Chaitanya K.; et al.
Macromolecules, 44(19), 7508-7510 (2011)
Investigating Cellular Structures at the Nanoscale with Organic Fluorophores.
van de Linde, S.; et al.
Chemistry and Biology, 20(1), 8-18 (2013)
Multiplexed 3D cellular super-resolution imaging with DNA-PAINT and Exchange-PAINT.
Jungmann, R.; Avendano, M.
Nature Methods, 11, 313-318 (2014)
2,2?-Thiodiethanol. A new water soluble mounting medium for high resolution optical microscopy.
Staudt, T.; et al.
Microscopy Research and Technique, 70(1), 1-9 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service