Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16426

Sigma-Aldrich

1,8-Diaminooctane 1 M solution

Synonym(s):

1,8-Diaminooctane solution, Additive Screening Solution 35/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

density

0.98 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

NCCCCCCCCN

InChI

1S/C8H20N2/c9-7-5-3-1-2-4-6-8-10/h1-10H2

InChI key

PWGJDPKCLMLPJW-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrizio Manetti et al.
Journal of medicinal chemistry, 52(15), 4774-4785 (2009-07-14)
Previous studies on agmatine and its derivatives suggested that the presence of hydrophobic groups on the guanidine moiety was a crucial key for inhibitory activity of maize polyamine oxidase. Accordingly, new lipophilic agmatine and iminoctadine derivatives were synthesized and tested
Richard A Hart et al.
Virology, 304(1), 97-104 (2002-12-20)
Proper function of the Rev regulatory system is essential for the replication of lentiviruses, including feline immunodeficiency virus (FIV) and human immunodeficiency virus type 1 (HIV-1). Specifically, Rev affects the overall stability of viral mRNAs that encode necessary structural and
R Yoda et al.
Biomaterials, 15(11), 944-949 (1994-09-01)
A-B-A type block copolymer (GIG(P)) membranes consisting of poly(N-hydroxypropyl-L-glutamine) (PHPG) as the A component and polyisoprene (PI) as the B component were prepared by carrying out aminoalcoholysis reaction with 3'-amino-1-propanol and a crosslinking reaction with 1,8-octamethylenediamine (OMDA) on membranes of
V Meza-Laguna et al.
Journal of nanoscience and nanotechnology, 7(10), 3563-3571 (2008-03-12)
We applied the direct solvent-free functionalization of fullerene C60 with aliphatic bifunctional amine, 1,8-diaminooctane, to prepare chemically cross-linked C60 thin films capable of binding silver nanoparticles. The gas-phase diamine treatment of C60 reduced dramatically the fullerene solubility in toluene, indicating
D J Fuller et al.
Radiation research, 95(1), 124-129 (1983-07-01)
The polyamines putrescine, spermidine, and spermine have been shown to sensitize Chinese hamster ovary (CHO) cells to the cytotoxic effects of elevated temperatures. This sensitization occurs in the order spermine greater than spermidine greater than putrescine. A series of homologs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service