Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16391

Sigma-Aldrich

Cellulose acetate sheets, humide

size 17.0 cm × 17.0 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NB.22

size

17.0 cm × 17.0 cm

Looking for similar products? Visit Product Comparison Guide

General description

After opening the package store membranes in 30% methanol. Do not let the membrane become dry. Before use store membranes for 10 minutes minimum in buffer. Apply sample on dull side. (Cut corner is bottom right!). Storage solution contains methanol.
After opening, store in 30% methanol. Do not let the membrane become dry. Before use, equilibrate membranes for at least 10 minutes in buffer. Apply sample on dull side (with the cut corner at bottom right). Storage solution contains methanol.

Application

For use in cellulose acetate electrophoresis

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service