Recommended Products
description
non-ionic
type
Type 15-S-5
Looking for similar products? Visit Product Comparison Guide
Other Notes
Polyglycol ether (nonionic) surfactants.
Legal Information
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
343.4 °F
flash_point_c
173 °C
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Dayna M Brichta-Harhay et al.
Applied and environmental microbiology, 74(20), 6289-6297 (2008-08-30)
The hide and carcass hygiene of cull cattle at slaughter in four geographically distant regions of the United States was examined from July 2005 to April 2006 by measuring the aerobic plate counts (APC) and the prevalences and loads of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service