Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15S30

Sigma-Aldrich

TERGITOL

Type 15-S-30

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900

description

non-ionic

type

Type 15-S-30

application(s)

detection

Looking for similar products? Visit Product Comparison Guide

Other Notes

Polyglycol ether (nonionic) surfactants.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

480.2 °F - closed cup

flash_point_c

249 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dayna M Brichta-Harhay et al.
Applied and environmental microbiology, 74(20), 6289-6297 (2008-08-30)
The hide and carcass hygiene of cull cattle at slaughter in four geographically distant regions of the United States was examined from July 2005 to April 2006 by measuring the aerobic plate counts (APC) and the prevalences and loads of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service