1522B
BIOEAZE™ Ethyl Vinyl Acetate (EVA) Bags
volume 350 mL, designed for use with Terumo Aseptic Connecting Device (ACD) Applications
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
sterility
sterile; γ-irradiated
packaging
pkg of 5 bags
volume
350 mL
Looking for similar products? Visit Product Comparison Guide
General description
Configured with:
Port P1: 1 Medicinal Port
Port P2: 1 x 1 ft length of opaque 001 C-Flex™ tubing with pinch clamp and plug
Port P3: 1 Spike Port
Port P1: 1 Medicinal Port
Port P2: 1 x 1 ft length of opaque 001 C-Flex™ tubing with pinch clamp and plug
Port P3: 1 Spike Port
Features and Benefits
BIOEAZE bioprocess bags are specifically designed to provide strong seals, extraordinary robustness, superior flex, crack and pin-hole resistance, plus desirable gas and moisture barrier performance.
BIOEAZE Ethyl Vinyl Acetate (EVA) bags are constructed from a single-web, medical grade laminate of four different films, with a fluid contact surface of EVA, with two layers of low-density polyethylene (LDPE) enveloping a layer of ethyl vinyl alcohol (EVOH).
BIOEAZE Ethyl Vinyl Acetate (EVA) bags are constructed from a single-web, medical grade laminate of four different films, with a fluid contact surface of EVA, with two layers of low-density polyethylene (LDPE) enveloping a layer of ethyl vinyl alcohol (EVOH).
Packaging
Individually Polybagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service