Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

146M-1

Sigma-Aldrich

CD45R (MB1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MB1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (146M-14)
vial of 0.5 mL concentrate (146M-15)
bottle of 1.0 mL predilute (146M-17)
vial of 1.0 mL concentrate (146M-16)
bottle of 7.0 mL predilute (146M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... PTPRC(5788)

General description

MB1 antibody stains preferentially B-cells and their neoplasms but is less specific as it will also react with some T-cell lymphomas and non-lymphoid tumors. The antigen for this antibody is in the membrane of all B-cells with the exception of plasma cells and some mature T-cells.

Quality


IVD

IVD

IVD

RUO

Linkage

CD45R Positive Control Slides, Product No. 146S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Poppema et al.
The American journal of pathology, 127(3), 418-429 (1987-06-01)
The absence of reactivity on routinely prepared tissue sections has hampered the use of monoclonal antileukocyte antibodies in diagnostic histopathology. Here we describe five new antibodies reactive with leukocyte subsets in formaldehyde-fixed, paraffin-embedded tissue sections. Antibody MT1 is reactive with
P S Master et al.
International journal of hematology, 55(3), 235-242 (1992-06-01)
The membrane expression of CD45R isoforms by leukaemic blasts from 92 cases of acute myeloid (AML) and 12 cases of lymphoblastic leukaemia was analysed by single and two-colour flow cytometry. Compared to normal mature granulocytes, which invariably expressed UCHL1 (CD45RO)
S W Chensue et al.
The American journal of pathology, 133(3), 564-572 (1988-12-01)
Using a highly specific rabbit antisera directed against murine tumor necrosis factor (TNF), immunohistochemical localization of this monokine was performed in cultured mouse peritoneal macrophages. Resident macrophages did not express TNF even after stimulus with lipopolysaccharide (LPS). In contrast, 12%
P A Hall et al.
Journal of clinical pathology, 40(8), 870-873 (1987-08-01)
A panel of antibodies recognising lymphoid and epithelial antigens in formalin fixed, paraffin embedded sections was applied to a series of 54 bone marrow trephines decalcified by formic or edetic acids. Normal trephines and cases infiltrated by myeloid, lymphoid, and
K P West et al.
The Journal of pathology, 150(2), 89-101 (1986-10-01)
The monoclonal antibodies MB1 and MT1, which detect B cells and T cells respectively, have been applied to human lymphoid tissues. The distribution of staining within paraffin sections was compared with that observed using frozen sections and was found to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service