Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14653

Sigma-Aldrich

3-[4-(Bromomethyl)phenyl]-7-(diethylamino)coumarin

BioReagent, suitable for fluorescence, ≥98.0% (N)

Synonym(s):

3-(4-(Bromomethyl)phenyl)-7-(diethylamino)-2H-1-benzopyran-2-one, MPAC-Br

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20BrNO2
CAS Number:
Molecular Weight:
386.28
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

assay

≥98.0% (N)

mp

150-155 °C

fluorescence

λex 402 nm; λem 476 nm in ethanol

suitability

suitable for fluorescence

SMILES string

CCN(CC)c1ccc2C=C(C(=O)Oc2c1)c3ccc(CBr)cc3

InChI

1S/C20H20BrNO2/c1-3-22(4-2)17-10-9-16-11-18(20(23)24-19(16)12-17)15-7-5-14(13-21)6-8-15/h5-12H,3-4,13H2,1-2H3

InChI key

WTCCANMBFQWZMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

HPLC-precolumn-derivatization reagent for carboxylic acids; improved "version" of the classical BMC (Cat. No. 17623), because it emits at longer wavelength and has stronger fluorescence. e.g. LOD of lauric acid: 17 fmol in 10 microliter[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Takechi et al.
Chemical & Pharmaceutical Bulletin, 44, 793-793 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service