14520
Bis(3-aminopropyl)amine
purum, ≥97.0% (GC)
Synonym(s):
3,3′-Diaminodipropylamine, 3,3′-Iminodipropylamine, N-(3-Aminopropyl)-1,3-propanediamine, Dipropylenetriamine, Norspermidine
About This Item
Recommended Products
grade
purum
assay
≥97.0% (GC)
refractive index
n20/D 1.481 (lit.)
n20/D 1.482
bp
151 °C/50 mmHg (lit.)
mp
−14 °C (lit.)
density
0.938 g/mL at 25 °C (lit.)
SMILES string
NCCCNCCCN
InChI
1S/C6H17N3/c7-3-1-5-9-6-2-4-8/h9H,1-8H2
InChI key
OTBHHUPVCYLGQO-UHFFFAOYSA-N
Gene Information
mouse ... Odc1(18263)
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Muta. 2 - Skin Corr. 1A - Skin Sens. 1A - STOT RE 2
target_organs
thymus
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 2
flash_point_f
242.6 °F - closed cup
flash_point_c
117 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service