Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14387

Sigma-Aldrich

L-Erythronic acid lithium salt

≥95% (TLC)

Synonym(s):

(2S,3S)-2,3,4-Trihydroxybutanoic acid lithium salt, Lithium L-erythronate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O5 · xLi+
CAS Number:
Molecular Weight:
136.10 (free acid basis)
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

assay

≥95% (TLC)

optical activity

[α]/D 34±4°, 24 hr, c = 0.1 in 1 M HCl

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@H](O)C(O)=O.[Li+]

InChI

1S/C4H8O5.Li/c5-1-2(6)3(7)4(8)9;/h2-3,5-7H,1H2,(H,8,9);/q;+1/t2-,3-;/m0./s1

InChI key

LXIXNEPXOOEPDK-GVOALSEPSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service