Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14149

Sigma-Aldrich

LUCY® 506

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500

fluorescence

λex 504 nm; λem 515 nm in aqueous buffer pH 8.3 with SDS and BSA

storage temp.

2-8°C

Application

Lucy-506 is a fluorescent stain for protein electrophoresis, with high sensitivity and easy, fast and robust staining procedure for all kinds of SDS gels. Protein staining by Lucy-506 does not interfere with subsequent MALDI-MS.
Sensitivity of routine procedure is ca. 3 ng/band, with linear response between 3 and 1000 ng/band

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

λmax. ~495 nm (MeOH)
Lucy-506 is provided as a 5000 × stock-solution in DMSO (5 mg/ml)

Legal Information

LUCY is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service