Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12670

Sigma-Aldrich

Tracy 645 (free acid)

suitable for fluorescence, ≥95.0% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500

assay

≥95.0% (HPCE)

solubility

DMF: soluble

fluorescence

λex 638 nm; λem 656 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

Tracy 645 is a newly developed NIR fluorescent Dye. The spectral range above 650 nm is of particular interested in biology analysis and imaging due to the reduced background and improved sensitivity.
Tracy 645 is highly water-soluble and stable dye.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service