Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12303C

Sigma-Aldrich

Fetal Bovine Serum

USDA approved countries, sterile-filtered, suitable for cell culture

Synonym(s):

FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
Pricing and availability is not currently available.

biological source

bovine fetus

sterility

sterile-filtered

quality

USDA approved countries

composition

Hemoglobin, ≤25 mg/dL

origin

non-USA origin

availability

not available in Japan

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Suitability

USDA Approved Sourcing refers to FBS produced from blood collected in countries outside of the United States that have been approved by the USDA (United States Department of Agriculture) for export of beef products into the United States. Eligible countries for export into the U.S. include Australia, Canada, Chile, Costa Rica, Honduras, Iceland, Japan, Mexico, New Zealand, Nicaragua, Uruguay. Foreign serum is tested to ensure the absence of exotic viruses before release by the USDA.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Legal Information

Available in USA, Canada, and Europe. Please check with your local supplier regarding availability and restrictions in other countries.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Katherine H Schreiber et al.
Nature communications, 10(1), 3194-3194 (2019-07-22)
Rapamycin, an inhibitor of mechanistic Target Of Rapamycin Complex 1 (mTORC1), extends lifespan and shows strong potential for the treatment of age-related diseases. However, rapamycin exerts metabolic and immunological side effects mediated by off-target inhibition of a second mTOR-containing complex
Yuzuru Itoh et al.
Science (New York, N.Y.), 371(6531), 846-849 (2021-02-20)
Mitochondrial ribosomes (mitoribosomes) are tethered to the mitochondrial inner membrane to facilitate the cotranslational membrane insertion of the synthesized proteins. We report cryo-electron microscopy structures of human mitoribosomes with nascent polypeptide, bound to the insertase oxidase assembly 1-like (OXA1L) through
Guadalupe Sepulveda et al.
eLife, 7 (2018-05-01)
As microtubule-organizing centers of animal cells, centrosomes guide the formation of the bipolar spindle that segregates chromosomes during mitosis. At mitosis onset, centrosomes maximize microtubule-organizing activity by rapidly expanding the pericentriolar material (PCM). This process is in part driven by
Abbie S Ireland et al.
Cancer cell, 38(1), 60-78 (2020-06-01)
Small cell lung cancer (SCLC) is a neuroendocrine tumor treated clinically as a single disease with poor outcomes. Distinct SCLC molecular subtypes have been defined based on expression of ASCL1, NEUROD1, POU2F3, or YAP1. Here, we use mouse and human
Perrine Barraud et al.
BMC neuroscience, 9, 24-24 (2008-02-20)
The olfactory epithelium is a neurogenic tissue comprising a population of olfactory receptor neurons that are renewed throughout adulthood by a population of stem and progenitor cells. Because of their relative accessibility compared to intra-cranially located neural stem/progenitor cells, olfactory

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Questions

1–2 of 2 Questions  
  1. Once a bottle of Fetal Bovine Serum (FBS) is thawed, for how long can the bottle be used?

    1 answer
    1. Once Fetal Bovine Serum has been thawed, the bottle can be used in cell culture for up to 7 days for most cell types when stored  refrigerated at 2-8°C.  If the cells are not fastidous (e.g. HeLa or fibroblasts), the thawed and refrigerated serum can be used for up to 30 days.  For some extremely fastidious cell lines, it may be necessary to screen individual lots for acceptable performance.  For uses such as blocking for various immuno procedures, usage will generally be for 30 days and perhaps somewhat longer when stored at 2-8°C.  For other procedures, usage beyond 30 days storage at 2-8°C will need to be determined.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service