Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12105C

Sigma-Aldrich

Fetal Bovine Serum

Dialyzed by ultrafiltration against 0.15M NaCl, USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
biological source:
bovine fetus
origin:
USA origin
sterility:
sterile-filtered
shipped in:
dry ice

biological source

bovine fetus

sterility

sterile-filtered

composition

Hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Preparation Note

10,000 Molecular Weight Cut-off

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruth M Risueño et al.
PloS one, 3(3), e1747-e1747 (2008-03-06)
How the T cell antigen receptor (TCR) discriminates between molecularly related peptide/Major Histocompatibility Complex (pMHC) ligands and converts this information into different possible signaling outcomes is still not understood. One current model proposes that strong pMHC ligands, but not weak
Li Zhou et al.
Genome biology, 9(7), R119-R119 (2008-07-31)
CD8+ NKT-like cells are naturally occurring but rare T cells that express both T cell and natural killer cell markers. These cells may play key roles in establishing tolerance to self-antigens; however, their mechanism of action and molecular profiles are
Silvia Goldoni et al.
The American journal of pathology, 173(3), 844-855 (2008-08-09)
Decorin, a member of the small leucine-rich proteoglycan gene family, down-regulates members of the ErbB receptor tyrosine kinase family and attenuates their signaling, leading to growth inhibition. We investigated the effects of decorin on the growth of ErbB2-overexpressing mammary carcinoma
Robert C Tuckey et al.
The Journal of steroid biochemistry and molecular biology, 112(4-5), 213-219 (2008-11-13)
Cytochrome P450scc (CYP11A1) metabolizes vitamin D3 to 20-hydroxyvitamin D3 as the major product, with subsequent production of dihydroxy and trihydroxy derivatives. The aim of this study was to determine whether cytochrome P450scc could metabolize 1alpha-hydroxyvitamin D3 and whether products were
Perrine Barraud et al.
BMC neuroscience, 9, 24-24 (2008-02-20)
The olfactory epithelium is a neurogenic tissue comprising a population of olfactory receptor neurons that are renewed throughout adulthood by a population of stem and progenitor cells. Because of their relative accessibility compared to intra-cranially located neural stem/progenitor cells, olfactory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service