12040
syn-Benzaldoxime
purum, ≥97.0% (GC)
Synonym(s):
(E)-Benzaldehyde oxime
About This Item
Recommended Products
grade
purum
Quality Level
assay
≥97.0% (GC)
form
solid
refractive index
n20/D 1.591 (lit.)
bp
104 °C/6 mmHg (lit.)
mp
34-36 °C (lit.)
solubility
methanol: 0.1 g/mL, clear
SMILES string
O\N=C\c1ccccc1
InChI
1S/C7H7NO/c9-8-6-7-4-2-1-3-5-7/h1-6,9H/b8-6+
InChI key
VTWKXBJHBHYJBI-SOFGYWHQSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
228.2 °F - closed cup
flash_point_c
109 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service