Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11520

Sigma-Aldrich

Azelaic acid dichloride

puriss., ≥98.5% (GC)

Synonym(s):

Azelaoyl chloride, Nonanedioyl chloride, Nonanedioyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClOC(CH2)7COCl
CAS Number:
Molecular Weight:
225.11
Beilstein/REAXYS Number:
1365424
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

assay

≥98.5% (GC)

refractive index

n20/D 1.467 (lit.)
n20/D 1.468

bp

166 °C/18 mmHg (lit.)

density

1.144 g/mL at 20 °C
1.143 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)CCCCCCCC(Cl)=O

InChI

1S/C9H14Cl2O2/c10-8(12)6-4-2-1-3-5-7-9(11)13/h1-7H2

InChI key

HGEVGSTXQGZPCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service